
Methyl-2-(aminosulphonylmethyl)benzoate ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAM1867-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:98.0 to 102.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:229.25 g/mol
EINECS:419-010-5
:GHS07
CAS Number:112941-26-1
Formula:C₉H₁₁NO₄S
MDL Number:MFCD03265370