Order Entry
Netherlands
ContactUsLinkComponent
Methyl-2-(aminosulphonyl)benzoate ≥98.0% (by total nitrogen basis)
Irritant
Methyl-2-(aminosulphonyl)benzoate ≥98.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
Irritant
Methyl-2-(aminosulphonyl)benzoate ≥98.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    124.0 to 129.0 deg-C

Specification Test Results

Frequently Bought Together