

2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine ≥98.0% (by HPLC, titration analysis)
TCIAM1865-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:163.0 to 167.0 deg-C
:
:154.17 g/mol
:401-360-5
:GHS07
:GHS08
:5248-39-5
:C₆H₁₀N₄O
:MFCD00585858