8-Methoxy-4-methylbenzo[g]coumarin ≥98.0% (by HPLC)
Catalog # TCIAM1862-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:218.0 to 223.0 deg-C
Specification Test Results
About this item
:
:240.26 g/mol
CAS Number:69169-71-7
Formula:C₁₅H₁₂O₃