
4-Methylthiazole-5-carbaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:82294-70-0
:
Formula:C₅H₅NOS
MDL Number:MFCD07368277
:127.161 g/mol
Boiling Pt:118°C
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:74.0 to 78.0 deg-C
Specification Test Results
Frequently Bought Together