Order Entry
Netherlands
ContactUsLinkComponent
N1-Methylbenzene-1,2-diamine dihydrochloride ≥98.0% (by HPLC, titration analysis)
 
N1-Methylbenzene-1,2-diamine dihydrochloride ≥98.0% (by HPLC, titration analysis)
  TCIAM1754-25G
 :  TCI
 :  
 
N1-Methylbenzene-1,2-diamine dihydrochloride ≥98.0% (by HPLC, titration analysis)
  TCIAM1754-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 98.0 %

 

 

 :
 :195.0900 g/mol
 :246-655-8
 :2811
 :6.1,III
 :GHS06
 :25148-68-9
 :C₇H₁₂Cl₂N₂
 :MFCD00042021