Mono-2-O-(p-toluenesulphonyl)-β-cyclodextrin Hydrate ≥97.0% (by HPLC)
TCIAM1741-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Specific rotation [a]20/D:+125.0 to +130.0 deg (C=4, DMSO)
- Water:max. 10.0 %
:
:1289.17 g/mol
:84216-71-7
:C₄₉H₇₆O₃₇S