Mono-6-O-(p-toluenesulphonyl)-γ-cyclodextrin ≥90.0% (by HPLC)
Catalog # TCIAM1645-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 90.0 area%
- Specific rotation [a]20/D:+146 to +151 deg (C=4, DMSO) (calcd.on anh.substance)
- Water:max. 15.0 %
Specification Test Results
About this item
:
:1451.31 g/mol
Storage Temperature:Refrigerator
CAS Number:97227-33-3
Formula:C₅₅H₈₆O₄₂S