4-Methoxyphenyl-3-O-allyl-2-azido-4,6-O-benzylidene-2-deoxy-β-D-galactopyranoside ≥95.0% (by HPLC)
TCIAM1643-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
:
:439.47 g/mol
:Freezer
:889453-83-2
:C₂₃H₂₅N₃O₆