4-Methoxyphenyl-3-O-allyl-2-azido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranoside ≥98.0% (by HPLC)
Catalog # TCIAM1638-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:119.0 to 123.0 deg-C
- Specific rotation [a]20/D:-30.0 to -32.5 deg (C=1, CHCl3)
- Elemental analysis (Nitrogen):9.10 to 10.10 %
Specification Test Results
About this item
:
:439.47 g/mol
Storage Temperature:Refrigerator
CAS Number:889453-78-5
Formula:C₂₃H₂₅N₃O₆
MDL Number:MFCD06797147