Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,1'-Methylenedi-2-naphthol ≥97.0% (by GC)
  TCIAM1410-25G
 :  TCI
 :  
 
1,1'-Methylenedi-2-naphthol ≥97.0% (by GC)
  TCIAM1410-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Brown powder to crystaline
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 95.0 %

 

 

 :
 :300.36 g/mol
 :214-145-4
 :GHS07
 :1096-84-0
 :C₂₁H₁₆O₂
 :MFCD00046470