Order Entry
Netherlands
ContactUsLinkComponent
Methyl-3-methyl-15-phenylpentadecanoate ≥95.0% (by GC)
Methyl-3-methyl-15-phenylpentadecanoate ≥95.0% (by GC)
Catalog # TCIAM1407-100MG
Supplier:  TCI
CAS Number:  
Methyl-3-methyl-15-phenylpentadecanoate ≥95.0% (by GC)
Catalog # TCIAM1407-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Green clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :346.56 g/mol
Boiling Pt:205°C
CAS Number:88336-88-3
Formula:C₂₃H₃₈O₂
MDL Number:MFCD04038423