Order Entry
Netherlands
ContactUsLinkComponent
1-(2-Mesitylene)-1,3-butanedione ≥96.0% (by GC)
1-(2-Mesitylene)-1,3-butanedione ≥96.0% (by GC)
Supplier:  TCI
Certificates
1-(2-Mesitylene)-1,3-butanedione ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White or Colorless to Yellow powder to lump to clear liquid
  • Purity (GC):
    min. 96.0 %
  • Melting point:
    43.0 to 47.0 deg-C
  • Solubility in Methanol:
    within almost transparency

Specification Test Results

Related Information

Frequently Bought Together