3-Mercapto-4-methyl-4H-1,2,4-triazole ≥98.0% (by HPLC, titration analysis)
Catalogus # TCIAM1224-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:165.0 to 169.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:115.15 g/mol
EINECS:246-496-4
:GHS07
CAS Number:24854-43-1
Formula:C₃H₅N₃S
MDL Number:MFCD00010069