
3-Mercapto-1-propanol ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:19721-22-3
:
Formula:C₃H₈OS
MDL Number:MFCD00192262
:92.16 g/mol
Boiling Pt:90°C
Flash Pt:85°C
UN:2810
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
- Purity (Iodometric Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together