

1-Methoxy-1-trimethylsilyloxypropene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:34880-70-1
:
Formula:C₇H₁₆O₂Si
MDL Number:MFCD00671554
:160.29 g/mol
Boiling Pt:52°C
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together