Order Entry
Netherlands
ContactUsLinkComponent
3-Methyl-4-cyclohexene-1,2-dicarboxylic anhydride ≥95.0% (by titrimetric analysis)
Corrosive
Health
3-Methyl-4-cyclohexene-1,2-dicarboxylic anhydride ≥95.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Corrosive
Health
3-Methyl-4-cyclohexene-1,2-dicarboxylic anhydride ≥95.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Morpholine Method):
    min. 95.0 %
  • Melting point:
    60.0 to 63.0 deg-C
  • Solubility in Methanol:
    within almost transparency

Specification Test Results

Frequently Bought Together