Order Entry
Netherlands
ContactUsLinkComponent
3-(Methylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
Flammable
Corrosive
3-(Methylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
Catalog # TCIAM1106-25G
Supplier:  TCI
CAS Number:  
Flammable
Corrosive
3-(Methylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
Catalog # TCIAM1106-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :100.17 g/mol
Boiling Pt:165°C
UN:2734
ADR:8,II
 :GHS02
 :GHS05
CAS Number:83030-08-4
Formula:C₅H₁₂N₂
MDL Number:MFCD00191460