
N-Methylmorpholine-N-oxide 50% ∼4.8 mol/L in water
Supplier: TCI
Certificates
About this item
CAS Number:7529-22-8
:
Formula:C₅H₁₁NO₂
MDL Number:MFCD00005947
:117.15 g/mol
EINECS:231-391-8
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Concentration (Nonaqueous Titration):49.0 to 55.0 w/w%
Specification Test Results
Frequently Bought Together