(S)-(+)-4-Methyl-1-hexanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1767-46-0
:
Formula:C₇H₁₆O
MDL Number:MFCD00221492
:116.20 g/mol
Boiling Pt:126°C
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Specific rotation [a]20/D:+6.0 to +9.0 deg (neat)
Specification Test Results
Frequently Bought Together