Order Entry
Netherlands
ContactUsLinkComponent
 
N,N'-Methylenebismethacrylamide ≥96.0% (by titrimetric analysis)
  TCIAM0930-25G
 :  TCI
 :  
N,N'-Methylenebismethacrylamide ≥96.0% (by titrimetric analysis)
  TCIAM0930-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Bromination Method):
    min. 96.0 %

 

 

 :
 :182.22 g/mol
 :219-102-3
 :2359-15-1
 :C₉H₁₄N₂O₂
 :MFCD00060096