Order Entry
Netherlands
ContactUsLinkComponent
2-Methyl-5-nitroimidazole-1-ethanol ≥99.0% (by GC, titration analysis)
Health
2-Methyl-5-nitroimidazole-1-ethanol ≥99.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Health
2-Methyl-5-nitroimidazole-1-ethanol ≥99.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Melting point:
    159.0 to 163.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together