

2-Methylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7003-32-9
:
Formula:C₇H₁₅N
MDL Number:MFCD00001492
:113.20 g/mol
Flash Pt:22°C
EINECS:230-277-5
UN:2733
ADR:3,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 % (total of isomers)
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
Frequently Bought Together