

4-Methoxy-4-methyl-2-pentanone ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:107-70-0
:
Formula:C₇H₁₄O₂
MDL Number:MFCD00043932
:130.19 g/mol
Boiling Pt:156°C
Flash Pt:48°C
EINECS:203-512-4
UN:2293
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together