Methyl propyl ether ≥95.0% (by GC)
Leverancier: TCI
Over dit artikel
Synoniemen: 1-Methoxypropane
- CAS-nummer:557-17-5
- Formule:C₄H₁₀O
- MDL-nummer:MFCD00059399
- Molecular Weight:74.12 g/mol
- Merck index:14,06113
- Kokend Pt:39°C
- EINECS:209-158-7
- UN:2612
- ADR:3,II
2 Opties Hieronder Beschikbaar
Product Details & Documents
Synoniemen: 1-Methoxypropane
- CAS-nummer:557-17-5
- Formule:C₄H₁₀O
- MDL-nummer:MFCD00059399
- Molecular Weight:74.12 g/mol
- Merck index:14,06113
- Kokend Pt:39°C
- EINECS:209-158-7
- UN:2612
- ADR:3,II
Specifications for Product Family
Specificatie testresultaten
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %



