5-Methyl-DL-tryptophan ≥98.0% (by HPLC, titration analysis)
: TCI
:951-55-3
:
:C₁₂H₁₄N₂O₂
:MFCD00005652
:218.26 g/mol
:213-453-6
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together