Leuco crystal violet ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:603-48-5
:
Formula:C₂₅H₃₁N₃
MDL Number:MFCD00008314
:373.54 g/mol
EINECS:210-043-9
:GHS07
Specification Test Results
- Appearance:White to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:177.0 to 181.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together