
1-Methylguanidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:21770-81-0
:22661-87-6
Formula:C₂H₈ClN₃
MDL Number:MFCD00012576
:109.557 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together