Order Entry
Netherlands
ContactUsLinkComponent
Methylene glycol bis(2,3-dibromopropyl)ether ≥70% (by GC)
Methylene glycol bis(2,3-dibromopropyl)ether ≥70% (by GC)
Catalog # TCIAM0313-25ML
Leverancier:  TCI
CAS-nummer:  
Methylene glycol bis(2,3-dibromopropyl)ether ≥70% (by GC)
Catalog # TCIAM0313-25ML
Leverancier:  TCI
CAS-nummer:  

Specificaties

  • Pk:
    25 mL
  • Seq:
    2315

Specificaties

Specificatie testresultaten

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 70.0 %

Specificatie testresultaten

Over dit artikel

 :
 :447.79 g/mol
Kokend Pt:220°C
CAS-nummer:34446-11-2
Formule:C₇H₁₂Br₄O₂
MDL-nummer:MFCD00059904