Order Entry
Netherlands
ContactUsLinkComponent
Methyl cellulose, 7,000-10,000 mPa.s (20 °C) (C=2%, H₂O)
Methyl cellulose, 7,000-10,000 mPa.s (20 °C) (C=2%, H₂O)
Supplier:  TCI
Certificates
Methyl cellulose, 7,000-10,000 mPa.s (20 °C) (C=2%, H₂O)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Viscosity:
    7,000 to 10,000 mPa-s (2 %, H2O, 20 deg-C)

Specification Test Results

Frequently Bought Together