Order Entry
Netherlands
ContactUsLinkComponent
 
 
N-Methyldi-N-octylamine ≥97.0% (by GC, titration analysis)
  TCIAM0232-25ML
 :  TCI
 :  
 
N-Methyldi-N-octylamine ≥97.0% (by GC, titration analysis)
  TCIAM0232-25ML
 :  TCI
 :  

 

  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %

 

 

 :
 :255.49 g/mol
 :303°C
 :118°C
 :224-703-9
 :2735
 :8,III
 :GHS05
 :4455-26-9
 :C₁₇H₃₇N
 :MFCD00009559