
L-778123 dihydrochloride ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:183499-56-1
:
Formula:C₂₂H₂₀ClN₅O·₂HCl
:478.80 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):13.20 to 15.40 %
- Chloride:14.1 to 15.4 %
Specification Test Results
Frequently Bought Together