Order Entry
Netherlands
ContactUsLinkComponent
Lithium diphenylphosphide ∼0.5 mol/L in THF
Flammable
Corrosive
Irritant
Health
Lithium diphenylphosphide ∼0.5 mol/L in THF
Catalog # TCIAL0302-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Lithium diphenylphosphide ∼0.5 mol/L in THF
Catalog # TCIAL0302-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Concentration (Nonaqueous Titration):
    9.5 to 11.3 w/w%

Specification Test Results

About this item

 :
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:4541-02-0
Formula:C₁₂H₁₀LiP
MDL Number:MFCD00046054