
Lithium dodecyl sulphate ≥98.0% (by titrimetric analysis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:2044-56-6
:
Formula:C₁₂H₂₅LiO₄S
MDL Number:MFCD00007467
:272.34 g/mol
EINECS:218-058-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization back titration):min. 98.0 %
- Suitability for protein-analysis:to pass test
Specification Test Results
Frequently Bought Together