L-Leucine ethyl ester hydrochloride ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2743-40-0
:
Formula:C₈H₁₇NO₂
MDL Number:MFCD00034879
:195.69 g/mol
EINECS:220-375-6
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 99.0 %
- Specific rotation [a]20/D:+18.0 to +20.0 deg (C=5, EtOH)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together