
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4511-42-6
:
Formula:C₆H₈O₄
MDL Number:MFCD00070594
:144.13 g/mol
EINECS:224-832-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:96.0 to 99.0 deg-C
- Specific rotation [a]20/D:-280 to -300 deg (C=1, Toluene)
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together