
Tetrapropylammonium hydroxide 10% in water for ion pair chromatography
Catalog # TCIAI1130-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):10.0 to 11.0 %
- Absorbance (230nm):max. 0.100
- Absorbance (254nm):max. 0.050
- Absorbance (400nm):max. 0.010
Specification Test Results
About this item
:
:203.37 g/mol
EINECS:224-800-6
UN:3267
ADR:8,III
:GHS05
CAS Number:4499-86-9
Formula:C₁₂H₂₉NO
MDL Number:MFCD00009360