
2-Iodo-1,3-phenylene bis(trifluoromethanesulphonate) ≥98.0% (by GC)
TCIAI1102-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:41.0 to 45.0 deg-C
:
:500.12 g/mol
:3261
:8,II
:GHS05
:514826-78-9
:C₈H₃F₆IO₆S₂