(6,6)Carbon Nanobelt bis(tetrahydrofuran) adduct ≥97.0% (by HPLC)
TCIAI1078-10MG
: TCI
:
- Pk:10 mg
- Seq:2275
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Carbon):88.00 to 90.50 %
:
:744.93 g/mol
:2245104-25-8
:C₄₈H₂₄·₂C₄H₈O