Order Entry
Netherlands
ContactUsLinkComponent
 
 
4-(1H-Imidazol-1-yl)benzoic acid ≥98.0% (by GC, titration analysis)
  TCIAI1017-1G
 :  TCI
 :  
 
4-(1H-Imidazol-1-yl)benzoic acid ≥98.0% (by GC, titration analysis)
  TCIAI1017-1G
 :  TCI
 :  

 

  • Pk:
    1 g/mol
  • Seq:
    9220

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

 

 

 :
 :188.19 g/mol
 :GHS07
 :17616-04-5
 :C₁₀H₈N₂O₂
 :MFCD00586645