
2-Isopropyl-2-methacryloyloxyadamantane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:297156-50-4
:
Formula:C₁₇H₂₆O₂
MDL Number:MFCD22571643
:262.39 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:38.0 to 42.0 deg-C
Specification Test Results
Frequently Bought Together