

3,3'-Iminobis(N,N-dimethylpropylamine) ≥97.0% (by GC, titration analysis)
Catalog # TCIAI0939-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
About this item
:
:187.33 g/mol
Flash Pt:98°C
EINECS:229-761-9
UN:2922
ADR:8,II
:GHS05
:GHS06
CAS Number:6711-48-4
Formula:C₁₀H₂₅N₃
MDL Number:MFCD00014880