5-Isopropyl-3,8-dimethylazulene-1-carbaldehyde ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3331-47-3
:
Formula:C₁₆H₁₈O
MDL Number:MFCD00275723
:226.32 g/mol
Melting Pt:83°C
Specification Test Results
- Appearance:Gray to Dark purple to Black powder to crystal
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together