

Imidapril hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:89396-94-1
:
Formula:C₂₀H₂₇N₃O₆
MDL Number:MFCD00923828
:441.91 g/mol
Merck Index:14,04911
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-65.0 to -69.0 deg (C=1, MeOH)
Specification Test Results
Frequently Bought Together