
N-Isopropylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:50632-53-6
:
Formula:C₃H₉NO
MDL Number:MFCD00012599
:111.57 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:69.0 to 73.0 deg-C
Specification Test Results
Frequently Bought Together