
4-Isopropylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Catalog # TCIAI0874-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
Boiling Pt:134°C
:170.25 g/mol
:GHS07
CAS Number:62067-45-2
Formula:C₁₀H₁₈O₂
MDL Number:MFCD01734696