Order Entry
Netherlands
ContactUsLinkComponent
Indirubin ≥97.0% (by HPLC)
Indirubin ≥97.0% (by HPLC)
Catalogus # TCIAI0868-25MG
Supplier:  TCI
CAS Number:  
undefined
Indirubin ≥97.0% (by HPLC)
Catalogus # TCIAI0868-25MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Red to Dark red to Dark purple powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Elemental analysis (Nitrogen):
    10.20 and le 11.00 %

Specification Test Results

About this item

 :
 :262.27 g/mol
Melting Pt:350°C
Storage Temperature:Refrigerator
CAS Number:479-41-4
Formula:C₁₆H₁₀N₂O₂
MDL Number:MFCD00221745