Order Entry
Netherlands
ContactUsLinkComponent
Isopimpinellin ≥98.0% (by GC)
Isopimpinellin ≥98.0% (by GC)
  TCIAI0861-10MG
 :  TCI
 :  
undefined
Isopimpinellin ≥98.0% (by GC)
  TCIAI0861-10MG
 :  TCI
 :  

 

  • Pk:
    10 mg
  • Seq:
    2275

 

 

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    148.0 to 152.0 deg-C

 

 

 :
 :246.22 g/mol
 :482-27-9
 :C₁₃H₁₀O₅
 :MFCD00017407