

4-Isopropylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Catalog # TCIAI0853-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:141.26 g/mol
Boiling Pt:90°C
UN:2733
ADR:3,III
:GHS02
:GHS05
CAS Number:52430-81-6
Formula:C₉H₁₉N
MDL Number:MFCD16853065