1-Isocyanoadamantane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:22110-53-8
:
Formula:C₁₁H₁₅N
MDL Number:MFCD02258889
:161.25 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together